Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTCAAAGGATGAAAATTAGTTG[A/C]TTTTTCTGTCTTAATTCTAGAAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf147 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C9orf147 - chromosome 9 open reading frame 147 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HSDL2 - hydroxysteroid dehydrogenase like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195822.1 | Intron | NP_001182751.1 | ||||
NM_032303.4 | Intron | NP_115679.2 | ||||
XM_011519091.2 | Intron | XP_011517393.1 | ||||
XM_017015203.1 | Intron | XP_016870692.1 | ||||
XM_017015204.1 | Intron | XP_016870693.1 | ||||
XM_017015205.1 | Intron | XP_016870694.1 |
Set Membership: |
HapMap |