Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605397 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD226 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CD226 - CD226 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303618.1 | 561 | Missense Mutation | NP_001290547.1 | |||
NM_001303619.1 | 561 | Missense Mutation | NP_001290548.1 | |||
NM_006566.3 | 561 | Missense Mutation | NP_006557.2 | |||
XM_005266642.3 | 561 | Missense Mutation | XP_005266699.1 | |||
XM_005266643.3 | 561 | Missense Mutation | XP_005266700.1 | |||
XM_006722374.3 | 561 | Missense Mutation | XP_006722437.1 | |||
XM_017025524.1 | 561 | Missense Mutation | XP_016881013.1 | |||
XM_017025525.1 | 561 | Missense Mutation | XP_016881014.1 | |||
XM_017025526.1 | 561 | Missense Mutation | XP_016881015.1 | |||
XM_017025527.1 | 561 | Missense Mutation | XP_016881016.1 |