Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCCCTAGGCCCTTTTGAGCTCT[A/C]ATATGTAGCATTATATAAAACCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615940 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HACD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HACD3 - 3-hydroxyacyl-CoA dehydratase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VWA9 - von Willebrand factor A domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136043.2 | Intron | NP_001129515.1 | ||||
NM_001207058.1 | Intron | NP_001193987.1 | ||||
NM_001207059.1 | Intron | NP_001193988.1 | ||||
XM_006720708.3 | Intron | XP_006720771.1 | ||||
XM_006720709.3 | Intron | XP_006720772.1 | ||||
XM_006720711.2 | Intron | XP_006720774.1 | ||||
XM_011522101.2 | Intron | XP_011520403.1 | ||||
XM_017022644.1 | Intron | XP_016878133.1 |