Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602887 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLG4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DLG4 - discs large MAGUK scaffold protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128827.2 | 2863 | UTR 3 | NP_001122299.1 | |||
NM_001321074.1 | 2863 | UTR 3 | NP_001308003.1 | |||
NM_001321075.1 | 2863 | UTR 3 | NP_001308004.1 | |||
NM_001321076.1 | 2863 | UTR 3 | NP_001308005.1 | |||
NM_001321077.1 | 2863 | UTR 3 | NP_001308006.1 | |||
NM_001365.4 | 2863 | UTR 3 | NP_001356.1 | |||
XM_005256491.1 | 2863 | UTR 3 | XP_005256548.1 | |||
XM_011523698.1 | 2863 | UTR 3 | XP_011522000.1 | |||
XM_011523699.2 | 2863 | UTR 3 | XP_011522001.1 | |||
XM_011523702.1 | 2863 | UTR 3 | XP_011522004.1 | |||
XM_017024288.1 | 2863 | UTR 3 | XP_016879777.1 | |||
XM_017024289.1 | 2863 | UTR 3 | XP_016879778.1 | |||
XM_017024290.1 | 2863 | UTR 3 | XP_016879779.1 |
Set Membership: |
Validated |