Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGCACTCCTTCTCTCATGGTGGT[C/T]CTGCCTTCATGACCTAATGTAAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607184 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC24B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SEC24B - SEC24 homolog B, COPII coat complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042734.3 | Intron | NP_001036199.1 | ||||
NM_001300813.2 | Intron | NP_001287742.1 | ||||
NM_001318085.1 | Intron | NP_001305014.1 | ||||
NM_001318086.1 | Intron | NP_001305015.1 | ||||
NM_006323.4 | Intron | NP_006314.2 | ||||
XM_005262688.1 | Intron | XP_005262745.1 | ||||
XM_005262691.1 | Intron | XP_005262748.1 | ||||
XM_011531535.2 | Intron | XP_011529837.1 | ||||
XM_011531536.2 | Intron | XP_011529838.1 | ||||
XM_011531537.2 | Intron | XP_011529839.1 | ||||
XM_011531539.2 | Intron | XP_011529841.1 | ||||
XM_011531540.2 | Intron | XP_011529842.1 | ||||
XM_017007652.1 | Intron | XP_016863141.1 |
Set Membership: |
HapMap |