Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAGTCACATCCCCAGAGTGCTC[C/T]CAGGTAAACAAGCCCCTGGGGGCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606030 MIM: 177015 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EDC4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EDC4 - enhancer of mRNA decapping 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRN1L - neuritin 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198443.1 | Intron | NP_940845.1 |
PSKH1 - protein serine kinase H1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |