Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGCTGGGGGTGAGGTTAGTGGAG[C/T]TGGGCAGATTGAAAGGCTAGATCTT
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 608845 MIM: 600066 | |||||||||||||||||||||||||||||
Literature Links: |
ARL6 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | ||||||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR - Not Available | ||||||||
AMR - Not Available |
ARL6 - ADP ribosylation factor like GTPase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006713783.3 | 871 | UTR 5 | XP_006713846.1 | |||
XM_017007311.1 | 871 | UTR 5 | XP_016862800.1 | |||
XM_017007312.1 | 871 | UTR 5 | XP_016862801.1 | |||
XM_017007313.1 | 871 | UTR 5 | XP_016862802.1 |
EPHA6 - EPH receptor A6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |