Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCCTTATCCATGTAAATGTTGT[C/T]CTAGTTGCAATTATTGCTCAAAACC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606152 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC19A3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
|||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
SLC19A3 - solute carrier family 19 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025243.3 | Intron | NP_079519.1 | ||||
XM_005246874.3 | Intron | XP_005246931.1 | ||||
XM_011511931.2 | Intron | XP_011510233.1 | ||||
XM_011511932.1 | Intron | XP_011510234.1 | ||||
XM_011511933.1 | Intron | XP_011510235.1 | ||||
XM_017005030.1 | Intron | XP_016860519.1 | ||||
XM_017005031.1 | Intron | XP_016860520.1 | ||||
XM_017005032.1 | Intron | XP_016860521.1 | ||||
XM_017005033.1 | Intron | XP_016860522.1 | ||||
XM_017005034.1 | Intron | XP_016860523.1 |
Set Membership: |
HapMap Validated |