Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTACCTTCCCTTGGGTTGCCAG[A/G]AGCAACAGCTTCATCTTGGGGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107941 MIM: 609455 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARRB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARRB2 - arrestin beta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257328.1 | Intron | NP_001244257.1 | ||||
NM_001257329.1 | Intron | NP_001244258.1 | ||||
NM_001257330.1 | Intron | NP_001244259.1 | ||||
NM_001257331.1 | Intron | NP_001244260.1 | ||||
NM_004313.3 | Intron | NP_004304.1 | ||||
NM_199004.1 | Intron | NP_945355.1 | ||||
XM_006721520.1 | Intron | XP_006721583.1 | ||||
XM_011523858.2 | Intron | XP_011522160.1 | ||||
XM_017024645.1 | Intron | XP_016880134.1 |
LOC101559451 - uncharacterized LOC101559451 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PELP1 - proline, glutamate and leucine rich protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |