Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAGATTTCTTCCTGAGTGGAAG[A/T]TCCTGACACACTTTCTGCCTCCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600058 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TXK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TXK - TXK tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003328.2 | Intron | NP_003319.2 | ||||
XM_011513747.2 | Intron | XP_011512049.1 | ||||
XM_011513748.2 | Intron | XP_011512050.1 | ||||
XM_017008580.1 | Intron | XP_016864069.1 | ||||
XM_017008581.1 | Intron | XP_016864070.1 | ||||
XM_017008582.1 | Intron | XP_016864071.1 |