Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACTGACAACTCACCAGTGAATTT[C/T]TGATTTACTTTCTGCCAGAATTTAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604439 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GAB1 - GRB2 associated binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002039.3 | Intron | NP_002030.2 | ||||
NM_207123.2 | Intron | NP_997006.1 | ||||
XM_006714167.3 | Intron | XP_006714230.1 | ||||
XM_006714168.3 | Intron | XP_006714231.1 | ||||
XM_017007966.1 | Intron | XP_016863455.1 | ||||
XM_017007967.1 | Intron | XP_016863456.1 | ||||
XM_017007968.1 | Intron | XP_016863457.1 | ||||
XM_017007969.1 | Intron | XP_016863458.1 | ||||
XM_017007970.1 | Intron | XP_016863459.1 | ||||
XM_017007971.1 | Intron | XP_016863460.1 |
MIR3139 - microRNA 3139 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |