Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGAGGAGTCAGATGGCTCCTTC[C/T]ACCTCATGCTAAGACCTAGGAACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603954 | ||||||||||||||||||||
Literature Links: |
CLDN25 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLDN25 - claudin 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101389.1 | 655 | Missense Mutation | CAC,TAC | H,Y 219 | NP_001094859.1 |
LOC390251 - SH3 domain containing GRB2 like 1 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZW10 - zw10 kinetochore protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |