Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606517 MIM: 601057 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHRR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AHRR - aryl-hydrocarbon receptor repressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242412.1 | Intron | NP_001229341.1 | ||||
NM_020731.4 | Intron | NP_065782.2 |
PDCD6 - programmed cell death 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267556.1 | Intron | NP_001254485.1 | ||||
NM_001267557.1 | Intron | NP_001254486.1 | ||||
NM_001267558.1 | Intron | NP_001254487.1 | ||||
NM_001267559.1 | Intron | NP_001254488.1 | ||||
NM_013232.3 | Intron | NP_037364.1 |
Set Membership: |
HapMap Validated |