Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAATACAAAAATAGAGAGATGGTG[C/T]CCGAGATTACCTGTGACGGTATTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605494 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EFCAB7 - EF-hand calcium binding domain 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032437.3 | Intron | NP_115813.2 | ||||
XM_005271274.3 | Intron | XP_005271331.1 | ||||
XM_006710976.3 | Intron | XP_006711039.1 | ||||
XM_006710977.1 | Intron | XP_006711040.1 | ||||
XM_011542301.2 | Intron | XP_011540603.1 | ||||
XM_017002548.1 | Intron | XP_016858037.1 |
ITGB3BP - integrin subunit beta 3 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206739.1 | Intron | NP_001193668.1 | ||||
NM_014288.4 | Intron | NP_055103.3 | ||||
XM_006710514.3 | Intron | XP_006710577.3 | ||||
XM_006710515.3 | Intron | XP_006710578.3 | ||||
XM_017000853.1 | Intron | XP_016856342.1 | ||||
XM_017000854.1 | Intron | XP_016856343.1 | ||||
XM_017000855.1 | Intron | XP_016856344.1 | ||||
XM_017000856.1 | Intron | XP_016856345.1 | ||||
XM_017000857.1 | Intron | XP_016856346.1 |
Set Membership: |
HapMap |