Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGTATTCGAGTGATTAGCGTTC[C/T]GGCTCCAGAACTGTCAAGAGGGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607719 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYT8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYT8 - synaptotagmin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290332.1 | Intron | NP_001277261.1 | ||||
NM_001290333.1 | Intron | NP_001277262.1 | ||||
NM_001290334.1 | Intron | NP_001277263.1 | ||||
NM_138567.4 | Intron | NP_612634.3 | ||||
XM_011520455.1 | Intron | XP_011518757.1 | ||||
XM_011520456.2 | Intron | XP_011518758.1 | ||||
XM_011520457.2 | Intron | XP_011518759.1 | ||||
XM_017018528.1 | Intron | XP_016874017.1 | ||||
XM_017018529.1 | Intron | XP_016874018.1 |