Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616129 MIM: 606822 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LURAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LURAP1 - leucine rich adaptor protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013615.2 | Intron | NP_001013633.1 |
POMGNT1 - protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243766.1 | Intron | NP_001230695.1 | ||||
NM_001290129.1 | Intron | NP_001277058.1 | ||||
NM_001290130.1 | Intron | NP_001277059.1 | ||||
NM_017739.3 | Intron | NP_060209.3 | ||||
XM_005271010.1 | Intron | XP_005271067.1 | ||||
XM_006710755.1 | Intron | XP_006710818.1 | ||||
XM_006710756.1 | Intron | XP_006710819.1 | ||||
XM_011541760.2 | Intron | XP_011540062.1 | ||||
XM_017001690.1 | Intron | XP_016857179.1 |
Set Membership: |
HapMap Validated |