Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAAATTCAGCCACAATAAAAGTG[C/T]GAATAAAGCAATATATCAATTGTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608283 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF21A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIF21A - kinesin family member 21A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173463.1 | 5616 | UTR 3 | NP_001166934.1 | |||
NM_001173464.1 | 5616 | UTR 3 | NP_001166935.1 | |||
NM_001173465.1 | 5616 | UTR 3 | NP_001166936.1 | |||
NM_017641.3 | 5616 | UTR 3 | NP_060111.2 | |||
XM_005269007.2 | 5616 | UTR 3 | XP_005269064.1 | |||
XM_005269008.2 | 5616 | UTR 3 | XP_005269065.1 | |||
XM_005269009.2 | 5616 | UTR 3 | XP_005269066.1 | |||
XM_005269010.2 | 5616 | UTR 3 | XP_005269067.1 | |||
XM_005269011.2 | 5616 | UTR 3 | XP_005269068.1 | |||
XM_005269012.2 | 5616 | UTR 3 | XP_005269069.1 | |||
XM_005269013.2 | 5616 | UTR 3 | XP_005269070.1 | |||
XM_005269014.2 | 5616 | UTR 3 | XP_005269071.1 | |||
XM_006719493.2 | 5616 | UTR 3 | XP_006719556.1 | |||
XM_006719494.2 | 5616 | UTR 3 | XP_006719557.1 | |||
XM_011538556.2 | 5616 | UTR 3 | XP_011536858.1 | |||
XM_017019607.1 | 5616 | UTR 3 | XP_016875096.1 | |||
XM_017019608.1 | 5616 | UTR 3 | XP_016875097.1 | |||
XM_017019609.1 | 5616 | UTR 3 | XP_016875098.1 | |||
XM_017019610.1 | 5616 | UTR 3 | XP_016875099.1 | |||
XM_017019611.1 | 5616 | UTR 3 | XP_016875100.1 |
Set Membership: |
HapMap |