Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATATACACTGACAGGAGCATTC[C/T]CTACTAAATTTCCCTATTTTCTGTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604043 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105748977 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC105748977 - uncharacterized LOC105748977 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEK2 - NIMA related kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204182.1 | Intron | NP_001191111.1 | ||||
NM_001204183.1 | Intron | NP_001191112.1 | ||||
NM_002497.3 | Intron | NP_002488.1 | ||||
XM_005273147.1 | Intron | XP_005273204.1 |
Set Membership: |
HapMap |