Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATAAATGTCCTTGACATCCTTT[A/G]CTCACGATAATGTTTTTGAGATTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602536 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB3GAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RAB3GAP1 - RAB3 GTPase activating protein catalytic subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172435.1 | Intron | NP_001165906.1 | ||||
NM_012233.2 | Intron | NP_036365.1 | ||||
XM_011510823.2 | Intron | XP_011509125.1 | ||||
XM_011510825.2 | Intron | XP_011509127.1 |
Set Membership: |
HapMap |