Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609531 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASGRP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RASGRP3 - RAS guanyl releasing protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139488.1 | Intron | NP_001132960.1 | ||||
NM_015376.2 | Intron | NP_056191.1 | ||||
NM_170672.2 | Intron | NP_733772.1 | ||||
XM_005264246.3 | Intron | XP_005264303.1 | ||||
XM_011532746.2 | Intron | XP_011531048.1 | ||||
XM_011532747.2 | Intron | XP_011531049.1 | ||||
XM_011532748.2 | Intron | XP_011531050.2 | ||||
XM_017003759.1 | Intron | XP_016859248.1 | ||||
XM_017003760.1 | Intron | XP_016859249.1 | ||||
XM_017003761.1 | Intron | XP_016859250.1 | ||||
XM_017003762.1 | Intron | XP_016859251.1 | ||||
XM_017003763.1 | Intron | XP_016859252.1 |
Set Membership: |
HapMap Validated |