Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKSG29 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FKSG29 - FKSG29 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR623 - microRNA 623 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBAC2 - UBA domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144072.1 | Intron | NP_001137544.1 | ||||
NM_177967.3 | Intron | NP_808882.1 | ||||
XM_006719948.3 | Intron | XP_006720011.1 | ||||
XM_011521082.2 | Intron | XP_011519384.1 | ||||
XM_011521083.2 | Intron | XP_011519385.1 | ||||
XM_011521084.2 | Intron | XP_011519386.1 | ||||
XM_017020553.1 | Intron | XP_016876042.1 |
Set Membership: |
HapMap Validated |