Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602359 MIM: 606684 MIM: 608303 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FXYD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FXYD1 - FXYD domain containing ion transport regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278717.1 | Intron | NP_001265646.1 | ||||
NM_001278718.1 | Intron | NP_001265647.1 | ||||
NM_005031.4 | Intron | NP_005022.2 | ||||
NM_021902.3 | Intron | NP_068702.1 | ||||
XM_017026874.1 | Intron | XP_016882363.1 | ||||
XM_017026875.1 | Intron | XP_016882364.1 | ||||
XM_017026876.1 | Intron | XP_016882365.1 |
FXYD7 - FXYD domain containing ion transport regulator 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022006.1 | Intron | NP_071289.1 |
LGI4 - leucine rich repeat LGI family member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |