Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCATTCAATTGAGGTTAAATCTT[C/T]ACACAAAGAAGGCATTAATAGCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601299 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMPR1A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BMPR1A - bone morphogenetic protein receptor type 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004329.2 | Intron | NP_004320.2 | ||||
XM_011540103.2 | Intron | XP_011538405.1 | ||||
XM_011540104.2 | Intron | XP_011538406.1 |
Set Membership: |
HapMap |