Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCATTTTCCTGGCTGCCCAGCCT[C/T]CATGTTACCATTATACATTTGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF37 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARHGEF37 - Rho guanine nucleotide exchange factor 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001669.2 | Intron | NP_001001669.2 | ||||
XM_005268447.4 | Intron | XP_005268504.1 | ||||
XM_005268448.1 | Intron | XP_005268505.1 | ||||
XM_006714784.3 | Intron | XP_006714847.1 | ||||
XM_011537642.2 | Intron | XP_011535944.1 | ||||
XM_017009460.1 | Intron | XP_016864949.1 | ||||
XM_017009461.1 | Intron | XP_016864950.1 |