Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611135 MIM: 607031 MIM: 603686 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KLB - klotho beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIAS - lipoic acid synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278590.1 | Intron | NP_001265519.1 | ||||
NM_001278591.1 | Intron | NP_001265520.1 | ||||
NM_001278592.1 | Intron | NP_001265521.1 | ||||
NM_006859.3 | Intron | NP_006850.2 | ||||
NM_194451.2 | Intron | NP_919433.1 | ||||
XM_006713990.3 | Intron | XP_006714053.1 | ||||
XM_017007665.1 | Intron | XP_016863154.1 | ||||
XM_017007666.1 | Intron | XP_016863155.1 | ||||
XM_017007667.1 | Intron | XP_016863156.1 |
RPL9 - ribosomal protein L9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000661.4 | Intron | NP_000652.2 | ||||
NM_001024921.3 | Intron | NP_001020092.1 |
Set Membership: |
HapMap |