Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612483 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAT3 - FAT atypical cadherin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008781.2 | Intron | NP_001008781.2 | ||||
XM_017017178.1 | Intron | XP_016872667.1 | ||||
XM_017017179.1 | Intron | XP_016872668.1 | ||||
XM_017017180.1 | Intron | XP_016872669.1 | ||||
XM_017017181.1 | Intron | XP_016872670.1 | ||||
XM_017017182.1 | Intron | XP_016872671.1 | ||||
XM_017017183.1 | Intron | XP_016872672.1 | ||||
XM_017017184.1 | Intron | XP_016872673.1 | ||||
XM_017017185.1 | Intron | XP_016872674.1 | ||||
XM_017017186.1 | Intron | XP_016872675.1 | ||||
XM_017017187.1 | Intron | XP_016872676.1 |
Set Membership: |
HapMap |