Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGGCCTTAGGTTCGGGCAGGTGT[A/G]GGGCCGCTCGCCCTCCGATGTTACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605847 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL7C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BCL7C - BCL tumor suppressor 7C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286526.1 | 1222 | UTR 3 | NP_001273455.1 | |||
NM_004765.3 | 1222 | Intron | NP_004756.2 | |||
XM_011545980.2 | 1222 | Intron | XP_011544282.1 | |||
XM_017023885.1 | 1222 | Intron | XP_016879374.1 | |||
XM_017023886.1 | 1222 | Intron | XP_016879375.1 | |||
XM_017023887.1 | 1222 | Intron | XP_016879376.1 | |||
XM_017023888.1 | 1222 | Intron | XP_016879377.1 |