Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609489 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MANBA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MANBA - mannosidase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005908.3 | 2380 | UTR 3 | NP_005899.3 | |||
XM_011531966.2 | 2380 | UTR 3 | XP_011530268.1 | |||
XM_017008203.1 | 2380 | UTR 3 | XP_016863692.1 | |||
XM_017008204.1 | 2380 | UTR 3 | XP_016863693.1 | |||
XM_017008205.1 | 2380 | UTR 3 | XP_016863694.1 |
Set Membership: |
HapMap |