Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATTATGTATTCAGCTTACCAGT[A/G]GTGCTTGTGGTATCTGTGGATTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDYHV1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
WDYHV1 - WDYHV motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283024.1 | Intron | NP_001269953.1 | ||||
NM_001283027.1 | Intron | NP_001269956.1 | ||||
NM_018024.2 | Intron | NP_060494.1 | ||||
XM_006716597.3 | Intron | XP_006716660.1 | ||||
XM_011517147.1 | Intron | XP_011515449.1 | ||||
XM_011517151.2 | Intron | XP_011515453.1 | ||||
XM_017013601.1 | Intron | XP_016869090.1 | ||||
XM_017013602.1 | Intron | XP_016869091.1 | ||||
XM_017013603.1 | Intron | XP_016869092.1 |