Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTCTGAGCTAGGCGCGAGGCCA[A/G]GGGGCTGCAGGAGTAGAAGACCAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610520 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD300LG PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CD300LG - CD300 molecule like family member g | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168322.1 | Intron | NP_001161794.1 | ||||
NM_001168323.1 | Intron | NP_001161795.1 | ||||
NM_001168324.1 | Intron | NP_001161796.1 | ||||
NM_145273.3 | Intron | NP_660316.2 | ||||
XM_005257075.2 | Intron | XP_005257132.1 | ||||
XM_005257076.2 | Intron | XP_005257133.1 | ||||
XM_006721721.2 | Intron | XP_006721784.1 | ||||
XM_006721722.2 | Intron | XP_006721785.1 | ||||
XM_011524384.2 | Intron | XP_011522686.1 | ||||
XM_017024235.1 | Intron | XP_016879724.1 |
Set Membership: |
HapMap |