Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604258 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DLC1 - DLC1 Rho GTPase activating protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164271.1 | 4148 | UTR 3 | NP_001157743.1 | |||
NM_001316668.1 | 4148 | UTR 3 | NP_001303597.1 | |||
NM_006094.4 | 4148 | UTR 3 | NP_006085.2 | |||
NM_024767.3 | 4148 | Intron | NP_079043.3 | |||
NM_182643.2 | 4148 | UTR 3 | NP_872584.2 | |||
XM_005273374.1 | 4148 | UTR 3 | XP_005273431.1 | |||
XM_017012951.1 | 4148 | UTR 3 | XP_016868440.1 | |||
XM_017012952.1 | 4148 | UTR 3 | XP_016868441.1 |
Set Membership: |
HapMap |