Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2CD2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C2CD2 - C2 calcium dependent domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015500.1 | 5210 | UTR 3 | NP_056315.1 | |||
NM_199050.2 | 5210 | UTR 3 | NP_950251.1 | |||
XM_005261109.4 | 5210 | Intron | XP_005261166.1 | |||
XM_011529522.1 | 5210 | UTR 3 | XP_011527824.1 | |||
XM_011529523.2 | 5210 | Intron | XP_011527825.1 | |||
XM_011529524.2 | 5210 | Intron | XP_011527826.1 | |||
XM_017028315.1 | 5210 | UTR 3 | XP_016883804.1 |
PRDM15 - PR domain 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |