Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAAATAATTGTCAAGTCAGGTG[A/G]CATACCTGAGGTTTCCTCTAAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602374 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNN3 - calponin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286055.1 | Intron | NP_001272984.1 | ||||
NM_001286056.1 | Intron | NP_001272985.1 | ||||
NM_001839.4 | Intron | NP_001830.1 | ||||
XM_017000245.1 | Intron | XP_016855734.1 |
SLC44A3 - solute carrier family 44 member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |