Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGTCTTTTCGCTGTCTCGATGGT[C/T]GATTCATTCTTTTTTTTTTATAATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616940 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXD2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
EXD2 - exonuclease 3'-5' domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193360.1 | Intron | NP_001180289.1 | ||||
NM_001193361.1 | Intron | NP_001180290.1 | ||||
NM_001193362.1 | Intron | NP_001180291.1 | ||||
NM_001193363.1 | Intron | NP_001180292.1 | ||||
NM_018199.3 | Intron | NP_060669.1 | ||||
XM_005267817.3 | Intron | XP_005267874.1 | ||||
XM_011536908.2 | Intron | XP_011535210.1 | ||||
XM_017021421.1 | Intron | XP_016876910.1 |
LOC105370550 - uncharacterized LOC105370550 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |