Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCAACAGCCGGAGCAGATCAGAC[C/T]GACCGGCCCTGCCCGCTCGGTCCCG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609247 | |||||||||||||||||||||||
Literature Links: |
RNF13 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
RNF13 - ring finger protein 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007282.4 | 100 | UTR 5 | NP_009213.1 | |||
NM_183381.2 | 100 | UTR 5 | NP_899237.1 | |||
XM_005247092.3 | 100 | UTR 5 | XP_005247149.1 | |||
XM_011512373.2 | 100 | UTR 5 | XP_011510675.1 | |||
XM_011512374.2 | 100 | UTR 5 | XP_011510676.1 | |||
XM_011512376.2 | 100 | Intron | XP_011510678.1 | |||
XM_017005654.1 | 100 | UTR 5 | XP_016861143.1 | |||
XM_017005655.1 | 100 | UTR 5 | XP_016861144.1 | |||
XM_017005656.1 | 100 | UTR 5 | XP_016861145.1 | |||
XM_017005657.1 | 100 | UTR 5 | XP_016861146.1 | |||
XM_017005658.1 | 100 | UTR 5 | XP_016861147.1 | |||
XM_017005659.1 | 100 | UTR 5 | XP_016861148.1 | |||
XM_017005660.1 | 100 | UTR 5 | XP_016861149.1 | |||
XM_017005661.1 | 100 | Intron | XP_016861150.1 |