Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTTGTGAGGTGAAACATGCACA[A/C]ATAAAGGGGGATGAGCACCAGCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
31 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600650 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf123 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C1orf123 - chromosome 1 open reading frame 123 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304759.1 | Intron | NP_001291688.1 | ||||
NM_001304760.1 | Intron | NP_001291689.1 | ||||
NM_017887.2 | Intron | NP_060357.1 |
CPT2 - carnitine palmitoyltransferase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |