Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
43 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616876 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC18 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCDC18 - coiled-coil domain containing 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306076.1 | Intron | NP_001293005.1 | ||||
NM_206886.4 | Intron | NP_996769.3 | ||||
XM_011541361.2 | Intron | XP_011539663.2 | ||||
XM_011541372.2 | Intron | XP_011539674.2 | ||||
XM_017001154.1 | Intron | XP_016856643.1 | ||||
XM_017001155.1 | Intron | XP_016856644.1 | ||||
XM_017001156.1 | Intron | XP_016856645.1 | ||||
XM_017001157.1 | Intron | XP_016856646.1 | ||||
XM_017001158.1 | Intron | XP_016856647.1 | ||||
XM_017001159.1 | Intron | XP_016856648.1 | ||||
XM_017001160.1 | Intron | XP_016856649.1 | ||||
XM_017001161.1 | Intron | XP_016856650.1 | ||||
XM_017001162.1 | Intron | XP_016856651.1 | ||||
XM_017001163.1 | Intron | XP_016856652.1 | ||||
XM_017001164.1 | Intron | XP_016856653.1 | ||||
XM_017001165.1 | Intron | XP_016856654.1 | ||||
XM_017001166.1 | Intron | XP_016856655.1 | ||||
XM_017001167.1 | Intron | XP_016856656.1 | ||||
XM_017001168.1 | Intron | XP_016856657.1 | ||||
XM_017001169.1 | Intron | XP_016856658.1 | ||||
XM_017001170.1 | Intron | XP_016856659.1 | ||||
XM_017001171.1 | Intron | XP_016856660.1 |
TMED5 - transmembrane p24 trafficking protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |