Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCCTGGCTCCACCAGGCTCCTTC[A/G]CTTCACAGTAGATTATGTCAGATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614968 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH2D4A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SH2D4A - SH2 domain containing 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174159.1 | Intron | NP_001167630.1 | ||||
NM_001174160.1 | Intron | NP_001167631.1 | ||||
NM_022071.3 | Intron | NP_071354.2 | ||||
XM_011544610.1 | Intron | XP_011542912.1 | ||||
XM_017013729.1 | Intron | XP_016869218.1 |
Set Membership: |
HapMap |