Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGAAGCCAGTGCCCCGGCATACG[C/T]CCTCTCGCGTCCCATTCCCCTCGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP85 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CEP85 - centrosomal protein 85 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281517.2 | Intron | NP_001268446.1 | ||||
NM_001281518.2 | Intron | NP_001268447.1 | ||||
NM_001319944.1 | Intron | NP_001306873.1 | ||||
NM_022778.4 | Intron | NP_073615.2 | ||||
XM_017002105.1 | Intron | XP_016857594.1 |
LOC101928303 - uncharacterized LOC101928303 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |