Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614543 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM69B PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
FAM69B - family with sequence similarity 69 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNHG7 - small nucleolar RNA host gene 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA17A - small nucleolar RNA, H/ACA box 17A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA17B - small nucleolar RNA, H/ACA box 17B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |