Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
45 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 170995 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ABCD3 - ATP binding cassette subfamily D member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122674.1 | Intron | NP_001116146.1 | ||||
NM_002858.3 | Intron | NP_002849.1 | ||||
XM_006710802.2 | Intron | XP_006710865.2 | ||||
XM_011541877.1 | Intron | XP_011540179.1 | ||||
XM_017001952.1 | Intron | XP_016857441.1 | ||||
XM_017001953.1 | Intron | XP_016857442.1 | ||||
XM_017001954.1 | Intron | XP_016857443.1 |
Set Membership: |
HapMap |