Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603760 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HUS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HUS1 - HUS1 checkpoint clamp component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SUN3 - Sad1 and UNC84 domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030019.1 | Intron | NP_001025190.1 | ||||
NM_001284350.1 | Intron | NP_001271279.1 | ||||
NM_152782.3 | Intron | NP_689995.3 | ||||
XM_017011930.1 | Intron | XP_016867419.1 |
Set Membership: |
HapMap Validated |