Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 238310 MIM: 611516 MIM: 165390 MIM: 600690 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AMT - aminomethyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NICN1 - nicolin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHOA - ras homolog family member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313941.1 | Intron | NP_001300870.1 | ||||
NM_001313943.1 | Intron | NP_001300872.1 | ||||
NM_001313944.1 | Intron | NP_001300873.1 | ||||
NM_001313945.1 | Intron | NP_001300874.1 | ||||
NM_001313946.1 | Intron | NP_001300875.1 | ||||
NM_001313947.1 | Intron | NP_001300876.1 | ||||
NM_001664.3 | Intron | NP_001655.1 |
TCTA - T-cell leukemia translocation altered | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022171.2 | Intron | NP_071503.1 |
Set Membership: |
HapMap Validated |