Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602944 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
E2F6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
E2F6 - E2F transcription factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278275.1 | 1144 | Intron | NP_001265204.1 | |||
NM_001278276.1 | 1144 | Intron | NP_001265205.1 | |||
NM_001278277.1 | 1144 | Intron | NP_001265206.1 | |||
NM_001278278.1 | 1144 | Intron | NP_001265207.1 | |||
NM_198256.3 | 1144 | Intron | NP_937987.2 | |||
NM_212540.2 | 1144 | Intron | NP_997705.1 | |||
XM_017003547.1 | 1144 | Missense Mutation | GGT,GTT | G,V 306 | XP_016859036.1 | |
XM_017003548.1 | 1144 | Missense Mutation | GGT,GTT | G,V 274 | XP_016859037.1 | |
XM_017003549.1 | 1144 | Missense Mutation | GGT,GTT | G,V 231 | XP_016859038.1 |