Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606004 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GGA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GGA1 - golgi associated, gamma adaptin ear containing, ARF binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001560.2 | Intron | NP_001001560.1 | ||||
NM_001172687.1 | Intron | NP_001166158.1 | ||||
NM_001172688.1 | Intron | NP_001166159.1 | ||||
NM_013365.4 | Intron | NP_037497.1 | ||||
XM_005261517.3 | Intron | XP_005261574.1 | ||||
XM_005261518.1 | Intron | XP_005261575.1 | ||||
XM_005261520.1 | Intron | XP_005261577.1 | ||||
XM_005261522.1 | Intron | XP_005261579.1 | ||||
XM_006724229.1 | Intron | XP_006724292.1 | ||||
XM_011530122.1 | Intron | XP_011528424.1 | ||||
XM_011530123.1 | Intron | XP_011528425.1 | ||||
XM_011530124.1 | Intron | XP_011528426.1 | ||||
XM_017028760.1 | Intron | XP_016884249.1 | ||||
XM_017028761.1 | Intron | XP_016884250.1 | ||||
XM_017028762.1 | Intron | XP_016884251.1 | ||||
XM_017028763.1 | Intron | XP_016884252.1 | ||||
XM_017028764.1 | Intron | XP_016884253.1 | ||||
XM_017028765.1 | Intron | XP_016884254.1 |