Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACGGCACTCCGTGGCAGCTTGA[G/A]GCCAGGAAAAGCAAGCCAGGGCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138249 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GRIN1 - glutamate ionotropic receptor NMDA type subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000832.6 | Intron | NP_000823.4 | ||||
NM_001185090.1 | Intron | NP_001172019.1 | ||||
NM_001185091.1 | Intron | NP_001172020.1 | ||||
NM_007327.3 | Intron | NP_015566.1 | ||||
NM_021569.3 | Intron | NP_067544.1 | ||||
XM_005266071.3 | Intron | XP_005266128.1 | ||||
XM_005266072.3 | Intron | XP_005266129.1 | ||||
XM_005266073.4 | Intron | XP_005266130.1 | ||||
XM_011518583.2 | Intron | XP_011516885.1 |
Set Membership: |
HapMap Validated |