Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCAGAGGCCCAGGAAGGAACCC[C/T]GAGCCCCCCACCAAAGCTAAAGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608083 MIM: 600745 MIM: 604783 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
APOC2 - apolipoprotein C2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
APOC4 - apolipoprotein C4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001646.2 | 147 | Missense Mutation | CCG,CTG | P,L 36 | NP_001637.1 |
APOC4-APOC2 - APOC4-APOC2 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CLPTM1 - CLPTM1, transmembrane protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |