Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109715 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSD3B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSD3B1 - hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000862.2 | 415 | Silent Mutation | GGA,GGT | G,G 90 | NP_000853.1 | |
XM_011541314.2 | 415 | Silent Mutation | GGA,GGT | G,G 92 | XP_011539616.1 | |
XM_017001119.1 | 415 | Silent Mutation | GGA,GGT | G,G 92 | XP_016856608.1 |
Set Membership: |
JSNP |