Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCAGGGACAGTTGGGGCCTACCT[C/G]GAGCAGCAGGCTAGGAAGAGGGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600533 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VANGL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
VANGL2 - VANGL planar cell polarity protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020335.2 | Intron | NP_065068.1 | ||||
XM_005245357.1 | Intron | XP_005245414.1 | ||||
XM_011509804.1 | Intron | XP_011508106.1 | ||||
XM_011509805.2 | Intron | XP_011508107.1 |