Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGCCCATTCTTTATCTGTGTGAG[A/G]CTTAGGGAGAGTAACTTCTTCACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611957 MIM: 608886 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR378A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR378A - microRNA 378a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPARGC1B - PPARG coactivator 1 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172698.1 | Intron | NP_001166169.1 | ||||
NM_001172699.1 | Intron | NP_001166170.1 | ||||
NM_133263.3 | Intron | NP_573570.3 | ||||
XM_005268372.4 | Intron | XP_005268429.1 | ||||
XM_011537553.2 | Intron | XP_011535855.1 | ||||
XM_011537554.2 | Intron | XP_011535856.1 | ||||
XM_011537555.2 | Intron | XP_011535857.1 | ||||
XM_011537556.2 | Intron | XP_011535858.1 | ||||
XM_011537557.1 | Intron | XP_011535859.1 |